Learn More
iQ Biosciences Purified Human CD4 T Cells, 10M cells/vial
Catalog No. NC2071960
Encompass_Preferred
High-quality, high-viability (> 90%) human CD4 T cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (typically > 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2071960 Supplier iQ Biosciences Supplier No. IQBHU1CD4T10
High-quality, high-viability (> 90%) human CD4 T cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (typically > 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.