Learn More
iQ Biosciences Purified Human B Cells, 10M cells/vial
Catalog No. NC2225345
Encompass_Preferred
High-quality, high-viability (> 90%) human B cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
This item is not returnable. View return policy
Catalog No. NC2225345 Supplier iQ Biosciences Supplier No. IQBHU1B10
High-quality, high-viability (> 90%) human B cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.