Learn More
iQ Biosciences Purified Human B Cells, 5M cells/vial
Catalog No. NC2855531
Encompass_Preferred
High-quality, high-viability (> 90%) human B cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2855531 Supplier iQ Biosciences Supplier No. IQBHU1B5
High-quality, high-viability (> 90%) human B cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.