Learn More
iQ Biosciences Purified Human CD3 Pan T Cells, 50M cells/vial
Supplier:  iQ Biosciences IQBHU1T50
Encompass
High-quality, high-viability (> 90%) human CD3 Pan T cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (> 99%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2855546