Learn More
iQ Biosciences Purified Human NK Cells, 5M cells/vial
Supplier:  iQ Biosciences IQBHU1NK5
Encompass
High-quality, high-viability (> 90%) human NK cells purified from PBMCs of healthy blood donors sourced from a world-renowned blood center. Highly enriched (typically > 90%) post-selection purity achieved via magnetic isolation. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2171690