Learn More
ATCC Purified; Human rhinovirus 16; Strain: 11757
Supplier:  ATCC VR283PQ
Encompass
Human rhinovirus 16 strain 11757 is a high-titer purified virus preparation that can be propagated in H1HeLa cells (ATCC CRL-1958). This strain was isolated in 1960 from the throat swab of a healthy 2-year-old human female in Washington, DC. It can be used in a variety of applications, including respiratory disease research, in vitro infectivity assays, antiviral studies, spiking studies, assay development, and vaccine development.
Catalog No. 50-238-2953