Learn More
ATCC Purified Human; rhinovirus 1B; Strain: B632
Supplier:  ATCC VR1645PQ
Encompass
Human rhinovirus 1B strain B632 is propagated in H1HeLa cells (ATCC CRL-1958). This purified preparation was isolated from human throat washings and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has a variety of applications, including respiratory disease research, assay development, vaccine development, and antiviral studies.
Catalog No. 50-238-2831