Learn More
ATCC Purified; Influenza A virus (H1N1); A/PR/8/34
Catalog No. 502383983
Encompass
Influenza A virus (H1N1) strain A/PR/8/34 is a high-titer, purified, live virus that is propagated in SPF chicken embryonated eggs. This strain was isolated in 1934 from a human patient in Puerto Rico and was deposited by the Centers for Disease Control and Prevention. This product has applications in respiratory disease research, antiviral studies, assay development, and vaccine development.
Catalog No. 50-238-3983 Supplier ATCC Supplier No. VR95PQ
Influenza A virus (H1N1) strain A/PR/8/34 is a high-titer, purified, live virus that is propagated in SPF chicken embryonated eggs. This strain was isolated in 1934 from a human patient in Puerto Rico and was deposited by the Centers for Disease Control and Prevention. This product has applications in respiratory disease research, antiviral studies, assay development, and vaccine development.