Learn More
Invivogen Purified Lipoteichoic acid from S. aureus (LTA-SA), 5mg
SDP
Catalog No. NC9694097
Encompass
Specificity: TLR2 agonist, Working concentration: 1 ng -1 μg/ml, Endotoxin level: EndoFit™, Lipoteichoic acid (LTA) is a major immunostimulatory component of Gram-positive bacteria. Like LPS, LTA is an amphiphile formed by a hydrophilic polyphosphate polymer linked to a neutral glycolipid. LTA stimulates immune cells through TLR2 to produce TNF-α and other inflammatory cytokines. Recognition of LTA involves also LPS-binding protein (LBP) and CD14.
Catalog No. NC9694097 Supplier Invivogen Supplier No. TLRLPSLTA
Specificity: TLR2 agonist, Working concentration: 1 ng -1 μg/ml, Endotoxin level: EndoFit™, Lipoteichoic acid (LTA) is a major immunostimulatory component of Gram-positive bacteria. Like LPS, LTA is an amphiphile formed by a hydrophilic polyphosphate polymer linked to a neutral glycolipid. LTA stimulates immune cells through TLR2 to produce TNF-α and other inflammatory cytokines. Recognition of LTA involves also LPS-binding protein (LBP) and CD14.