Learn More
Cayman Chemical Purified Mixed Gangliosides (porcine) (ammonium salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502933924
Encompass_Preferred
A mixture of purified porcine gangliosides; found in all cells but at particularly high levels in the brain; typically contain stearic acid as the fatty acyl chain; modulate intra- and inter-cell signaling and regulate receptor function; involved in developmental processes; implicated in tumorigenesis
Catalog No. 50-293-3924 Supplier Cayman Chemical Supplier No. 248571 MG
A mixture of purified porcine gangliosides; found in all cells but at particularly high levels in the brain; typically contain stearic acid as the fatty acyl chain; modulate intra- and inter-cell signaling and regulate receptor function; involved in developmental processes; implicated in tumorigenesis