Learn More
Cayman Chemical PurIfIed MIxed GnglIosIdS 25mg
SDP
Catalog No. 502933943
Encompass_Preferred
A mixture of purified porcine gangliosides; found in all cells but at particularly high levels in the brain; typically contain stearic acid as the fatty acyl chain; modulate intra- and inter-cell signaling and regulate receptor function; involved in developmental processes; implicated in tumorigenesis
Catalog No. 50-293-3943 Supplier Cayman Chemical Supplier No. 2485725 MG
A mixture of purified porcine gangliosides; found in all cells but at particularly high levels in the brain; typically contain stearic acid as the fatty acyl chain; modulate intra- and inter-cell signaling and regulate receptor function; involved in developmental processes; implicated in tumorigenesis