Learn More
Chondrex Inc PURIFIED OVA, LOW ENDOTOXIN
Catalog No. 501526751
Encompass_Preferred
10 mg (lyophilized) of ovalbumin purified from chicken egg white. >98% pure by agarose fel analysis. Endotoxin level: < 1 EU/mg. Intended for use as antigen in airway hyperresponsiveness models.
Catalog No. 50-152-6751 Supplier Chondrex Inc Supplier No. 3022
10 mg (lyophilized) of ovalbumin purified from chicken egg white. >98% pure by agarose fel analysis. Endotoxin level: < 1 EU/mg. Intended for use as antigen in airway hyperresponsiveness models.