Learn More
OriGene RIP (RIPK1) (NM_003804) Human Recombinant Protein, 50 ug
SDP
Supplier:  OriGene TP760433
Encompass_Preferred
Purified recombinant protein of Human receptor (TNFRSF)-interacting serine-threonine kinase 1 (RIPK1), full length, with N-terminal HIS tag, expressed in E.Coli, 50ug.
Catalog No. NC1191472