Learn More
iQ Biosciences Purified Rhesus Monkey CD14+ Cells, 2M cells/vial
Supplier:  iQ Biosciences IQBMN2CD142
Encompass_Preferred
High-quality, high-viability (> 90%) CD14+ cells purified from rhesus macaque peripheral blood mononuclear cells (PBMC) by magnetic isolation. Highly enriched (typically > 90%) post-selection purity achieved via magnetic isolation. Ideal source for rhesus monocytes and neutrophils. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available:- Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor- Donor-matched PBMCs
Catalog No. NC2143114