Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Purified Viral Lysate, Epstein-Barr Virus (EBV)
SDP

Catalog No. 22157397 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-397 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-397 Supplier ZeptoMetrix Supplier No. 0810008
Add to Cart
Add to Cart

Can be utilized as antigen, source for purification of viral proteins or for detection of viral antibodies

  • Concentration ranges from 0.5 to 3.0mg/mL
  • Available in 1mg vials

Immunoassays, Western Blot, Dot Blot and other protein-based assays

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration 0.5 to 3mg/mL
Content And Storage -65°C
Detects Viral Antibodies
Form Solution
Media Type Vial
Product Type Purified Viral Lysate
For Use With (Application) Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay
Purification Method Sucrose density gradient ultracentrifugation
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.