Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Purified Viral Lysate, Influenza A H1N1 (California/07/09) (Swine Flu)
SDP

Catalog No. 22157476 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-476 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-476 Supplier ZeptoMetrix Supplier No. 0810165
Add to Cart
Add to Cart

Can be utilized as an antigen, as a source for the purification of viral proteins, or for the detection of viral antibodies

  • Propagated in the MDCK cell line
  • Purified using sucrose density gradient ultracentrifugation, disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6M KCl, and heat inactivated

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration 0.5 to 3mg/mL
Content And Storage -65°C
Detects Viral Antibodies
Form Solution
Media Type Vial
Product Type Purified Viral Lysate
For Use With (Application) Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay
Purification Method Sucrose density gradient ultracentrifugation
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.