Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Purified Viral Lysate, Influenza A H3N2 (HK/8/68)
SDP

Catalog No. 22157940 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mg/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-940 1 mg/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-940 Supplier ZeptoMetrix Supplier No. 0810250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be utilized as an antigen, as a source for the purification of viral proteins, or for the detection of viral antibodies

  • Propagated in the MDCK cell line
  • Purified using sucrose density gradient ultracentrifugation, disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6M KCl, and heat inactivated

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Concentration 0.5 to 3mg/mL
Content And Storage -65°C or below
Form Solution
Product Type Influenza A H3N2 Virus Infectious Culture Fluid
For Use With (Application) Immunodetection of antibodies to Influenza virus using solid-phase EIA, Western blot, Dot blot
Packaging Type Vial
Purification Method Sucrose density gradient ultracentrifugation
Quantity 1 mg/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.