Learn More
Description
Purified Water USP is a clear, colorless, odorless liquid.
Not for parenteral administration. Not for use in sterile irrigation, injection, infusion, or as potable product. Intended only for manufacturing and general laboratory use.
Specifications
Specifications
| Chemical Name or Material | Purified Water USP |
| Packaging | White High Gloss Polypropylene 10 in.W x 4.17 in.H - 7.6 in.OD |
| CAS | 7732-18-5 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 18.02 |
| Quantity | 3.8 L |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
