Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Purified Water USP
SDP

Catalog No. 14185526 Shop All Medisca Products
Change view
Click to view available options
Quantity:
3.8 L
19 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-526 19 L
14-185-469 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-185-526 Supplier Medisca Supplier No. 094607
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purified Water USP is a clear, colorless, odorless liquid. Not for parenteral administration.

Purified Water USP is a clear, colorless, odorless liquid.

Not for parenteral administration. Not for use in sterile irrigation, injection, infusion, or as potable product. Intended only for manufacturing and general laboratory use.

Specifications

Chemical Name or Material Purified Water USP
Packaging White High Gloss Polypropylene 10 in.W x 4.17 in.H - 7.6 in.OD
CAS 7732-18-5
Type Drug Standard
Molecular Weight (g/mol) 18.02
Quantity 19 L
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.