Learn More
ATCC Purified; Human respiratory; syncytial virus; Strain: 18537
Catalog No. 502382387
Encompass
Human respiratory syncytial virus strain 18537 is a high-titer preparation that is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in 1962 from the respiratory secretions of a child with acute respiratory disease seen at Children’s Hospital in Washington, DC, and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in virucide testing, respiratory research, assay development and vaccine development.
Catalog No. 50-238-2387 Supplier ATCC Supplier No. VR1580PQ
Human respiratory syncytial virus strain 18537 is a high-titer preparation that is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in 1962 from the respiratory secretions of a child with acute respiratory disease seen at Children’s Hospital in Washington, DC, and was deposited by the National Institute of Allergy and Infectious Diseases (NIAID). It has applications in virucide testing, respiratory research, assay development and vaccine development.