Learn More
Vici Metronics Liquid Carbon Dioxide Purifier, 1/8'' Fitting Size, 22.5'' Length
SDP
Catalog No. NC0485613
Encompass_Preferred
Produce SFC/SFE grade CO2 from Coleman grade CO2. Remove oxygen, moisture, sulfur compounds, halocarbons, and most hydrocarbons.
Catalog No. NC0485613 Supplier Vici Metronics Supplier No. P7001
Produce SFC/SFE grade CO2 from Coleman grade CO2. Remove oxygen, moisture, sulfur compounds, halocarbons, and most hydrocarbons.