Learn More
Advion Inc PURIFLASH C18-XS 15UM F0012 FLASH COLUMN (Pack of 2)
SDP
Catalog No. NC3793933
Encompass
Ultra-Pur PSDVB(Atoll X)100 Å – 800 m2/g40 μmpH stability: 1.0 to 13.0A Universal polymer with a largespecific surface for the purification ofmedium and non-polar compoundswith Mw < 5 KD under pHconditions from 1 to 13.
Catalog No. NC3793933 Supplier Advion Inc Supplier No. PFXF0012
Ultra-Pur PSDVB(Atoll X)100 Å – 800 m2/g40 μmpH stability: 1.0 to 13.0A Universal polymer with a largespecific surface for the purification ofmedium and non-polar compoundswith Mw < 5 KD under pHconditions from 1 to 13.