Learn More
Advion Inc PURIFLASH® • X • 40UM • FLASH COLUMN, 40UM F0004 FLASH COLUMN, 4g, 4/PK
SDP
Catalog No. NC3793932
Encompass
Ultra-Pur PSDVB(Atoll X)100 Å – 800 m2/g40 μmpH stability: 1.0 to 13.0A Universal polymer with a largespecific surface for the purification ofmedium and non-polar compoundswith Mw < 5 KD under pHconditions from 1 to 13.
Catalog No. NC3793932 Supplier Advion Inc Supplier No. PFXF0004
Ultra-Pur PSDVB(Atoll X)100 Å – 800 m2/g40 μmpH stability: 1.0 to 13.0A Universal polymer with a largespecific surface for the purification ofmedium and non-polar compoundswith Mw < 5 KD under pHconditions from 1 to 13.