Learn More
Medchemexpress LLC Purine | 120-73-0 | 99.8% | 120.11 | 1 ML
SDP
Catalog No. 5000404055
Encompass_Preferred
Purine is an endogenous metabolite whose bases are the building blocks of nucleic acids. It protects against oxidant-induced cell injury by inhibiting the activation of PARP, making it suitable for research into cancer and nervous system diseases.
  • Inhibits PARP activation
  • Protects against oxidant-induced cell injury
  • Building block of nucleic acids
  • Supports research in cancer and nervous system diseases
Catalog No. 50-004-04055 Supplier Medchemexpress LLC Supplier No. HY3443110MM/1ML
Purine is an endogenous metabolite whose bases are the building blocks of nucleic acids. It protects against oxidant-induced cell injury by inhibiting the activation of PARP, making it suitable for research into cancer and nervous system diseases.
  • Inhibits PARP activation
  • Protects against oxidant-induced cell injury
  • Building block of nucleic acids
  • Supports research in cancer and nervous system diseases