Learn More
Selleck Chemical LLC Purine
SDP
Catalog No. 5000002950
Encompass_Preferred
Purine is a heterocyclic aromatic organic compound which plays crucial roles in DNA and RNA It is also significant components in a number of other important biomolecules such as ATP GTP cyclic AMP NADH and coenzyme A
Catalog No. 50-000-02950 Supplier Selleck Chemical LLC Supplier No. S635925MG
Purine is a heterocyclic aromatic organic compound which plays crucial roles in DNA and RNA It is also significant components in a number of other important biomolecules such as ATP GTP cyclic AMP NADH and coenzyme A