Learn More
Apexbio Technology LLC PURINE-200MG
SDP
Catalog No. 5000568630
Encompass_Preferred
Purine (CAS No 120-73-0) is a key compound in biomedical research originating from a class of heterocyclic aromatic organic compounds It is integral to numerous biological processes functioning primarily through its role in nucleic acid synthesis targeting nucleotides and pathways involved in DNA and RNA metabolism Purine s in vitro activity often falls within the nanomolar to low micromolar range making it highly relevant for studies on cell proliferation and signal transduction It is commonly utilized in cell models animal studies and drug screening with particular emphasis on understanding metabolic pathways and evaluating antimetabolite therapies Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68630 Supplier Apexbio Technology LLC Supplier No. C6588200
Purine (CAS No 120-73-0) is a key compound in biomedical research originating from a class of heterocyclic aromatic organic compounds It is integral to numerous biological processes functioning primarily through its role in nucleic acid synthesis targeting nucleotides and pathways involved in DNA and RNA metabolism Purine s in vitro activity often falls within the nanomolar to low micromolar range making it highly relevant for studies on cell proliferation and signal transduction It is commonly utilized in cell models animal studies and drug screening with particular emphasis on understanding metabolic pathways and evaluating antimetabolite therapies Experimental concentrations typically depend on the specific research design