Learn More
Medchemexpress LLC Purine nucleoside phosphorylase, Microorganism | 100 U
SDP
Catalog No. 5000400388
Encompass_Preferred
Purine nucleoside phosphorylase, Microorganism (PNP) is a key enzyme in purine metabolism. It is involved in the purine rescue pathway, and its deficiency results in impaired T-cell function. In the presence of inorganic orthophosphate as the second substrate, Purine nucleoside phosphorylase catalyzes the breaking of the glycosidic bond between ribose and deoxyribonucleoside to generate purine base and ribose (deoxyribose)-1-phosphate.
  • Key enzyme in purine metabolism
  • Involved in the purine rescue pathway
  • Deficiency impairs T-cell function
  • Catalyzes the breaking of glycosidic bonds
  • Generates purine base and ribose (deoxyribose)-1-phosphate
  • Target: endogenous metabolite
  • Research areas: metabolic disease
Catalog No. 50-004-00388 Supplier Medchemexpress LLC Supplier No. HYP2724100U
Purine nucleoside phosphorylase, Microorganism (PNP) is a key enzyme in purine metabolism. It is involved in the purine rescue pathway, and its deficiency results in impaired T-cell function. In the presence of inorganic orthophosphate as the second substrate, Purine nucleoside phosphorylase catalyzes the breaking of the glycosidic bond between ribose and deoxyribonucleoside to generate purine base and ribose (deoxyribose)-1-phosphate.
  • Key enzyme in purine metabolism
  • Involved in the purine rescue pathway
  • Deficiency impairs T-cell function
  • Catalyzes the breaking of glycosidic bonds
  • Generates purine base and ribose (deoxyribose)-1-phosphate
  • Target: endogenous metabolite
  • Research areas: metabolic disease