Learn More
Medchemexpress LLC Purinostat mesylate | 2650188-32-0 | 99.45% | 586.62 | 100 MG
SDP
Catalog No. 5000361751
Encompass_Preferred
Purinostat mesylate is a selective inhibitor of HDAC, inhibiting class I and class IIb HDACs. It induces apoptosis, affects the cell cycle, and exhibits potent anti-leukemia effects in vivo, making it suitable for research on lymphoblastic leukemia.
  • Selective inhibitor of HDACs
  • Inhibits class I and class IIb HDACs (IC50s 0.81 to 11.5 nM)
  • Induces apoptosis
  • Affects cell cycle of LAMA84 and 188 BL-2 cells
  • Exhibits potent anti-leukemia effects in vivo
  • Suitable for research on lymphoblastic leukemia
  • High purity: 99.45%
Catalog No. 50-003-61751 Supplier Medchemexpress LLC Supplier No. HY150109100MG
Purinostat mesylate is a selective inhibitor of HDAC, inhibiting class I and class IIb HDACs. It induces apoptosis, affects the cell cycle, and exhibits potent anti-leukemia effects in vivo, making it suitable for research on lymphoblastic leukemia.
  • Selective inhibitor of HDACs
  • Inhibits class I and class IIb HDACs (IC50s 0.81 to 11.5 nM)
  • Induces apoptosis
  • Affects cell cycle of LAMA84 and 188 BL-2 cells
  • Exhibits potent anti-leukemia effects in vivo
  • Suitable for research on lymphoblastic leukemia
  • High purity: 99.45%