Learn More
Medchemexpress LLC Purinostat mesylate | 2650188-32-0 | 99.5% | 586.62 | 25 MG
SDP
Catalog No. 5000360005
Encompass_Preferred
Purinostat mesylate is a selective inhibitor of HDAC. It inhibits class I and class IIb HDACs with IC50s ranging from 0.81 to 11.5 nM. It induces apoptosis and affects the cell cycle of LAMA84 and 188 BL-2 cells, showing potent anti-leukemia effects in vivo. It is used for research on lymphoblastic leukemia.
  • Inhibits class I and class IIb HDACs
  • Induces apoptosis and affects cell cycle of LAMA84 and 188 BL-2 cells
  • Suppresses leukemia progression in vivo
  • Demonstrates potent anti-leukemia effects in BCR-ABL(T315I)-induced primary B-ALL mice
Catalog No. 50-003-60005 Supplier Medchemexpress LLC Supplier No. 25MGHY150109
Purinostat mesylate is a selective inhibitor of HDAC. It inhibits class I and class IIb HDACs with IC50s ranging from 0.81 to 11.5 nM. It induces apoptosis and affects the cell cycle of LAMA84 and 188 BL-2 cells, showing potent anti-leukemia effects in vivo. It is used for research on lymphoblastic leukemia.
  • Inhibits class I and class IIb HDACs
  • Induces apoptosis and affects cell cycle of LAMA84 and 188 BL-2 cells
  • Suppresses leukemia progression in vivo
  • Demonstrates potent anti-leukemia effects in BCR-ABL(T315I)-induced primary B-ALL mice