Learn More
Medchemexpress LLC Purinostat mesylate | 2650188-32-0 | 99.5% | 586.62 | 5 MG
SDP
Catalog No. 5000361752
Encompass_Preferred
Purinostat mesylate is a selective inhibitor of HDAC, inhibiting class I and class IIb HDACs with IC50s ranging from 0.81 to 11.5 nM. It induces apoptosis and affects the cell cycle, demonstrating potent anti-leukemia effects in vivo. This product can be used for research on lymphoblastic leukemia.
  • Selective inhibitor of HDAC
  • Inhibits class I and class IIb HDACs
  • Induces apoptosis
  • Affects the cell cycle of LAMA84 and 188 BL-2 cells
  • Demonstrates potent anti-leukemia effects in vivo
  • Can be used for research on lymphoblastic leukemia
Catalog No. 50-003-61752 Supplier Medchemexpress LLC Supplier No. HY1501095MG
Add to Cart
Add to Cart
Purinostat mesylate is a selective inhibitor of HDAC, inhibiting class I and class IIb HDACs with IC50s ranging from 0.81 to 11.5 nM. It induces apoptosis and affects the cell cycle, demonstrating potent anti-leukemia effects in vivo. This product can be used for research on lymphoblastic leukemia.
  • Selective inhibitor of HDAC
  • Inhibits class I and class IIb HDACs
  • Induces apoptosis
  • Affects the cell cycle of LAMA84 and 188 BL-2 cells
  • Demonstrates potent anti-leukemia effects in vivo
  • Can be used for research on lymphoblastic leukemia