Learn More
Medchemexpress LLC Purinostat mesylate | 2650188-32-0 | 99.5% | 586.62 g/mol | C24H30N10O6S | 10 MG
SDP
Catalog No. 5000286225
Encompass_Preferred
Purinostat mesylate is the mesylate salt of purinostat, a potent and selective histone deacetylase (HDAC) inhibitor used in preclinical research. It exhibits low-nanomolar potency against multiple HDAC isoforms and has demonstrated apoptosis induction and cell-cycle effects in cancer models. The product is supplied as a solid or as a ready-to-use DMSO solution for laboratory studies.
  • Potent HDAC inhibition at low-nanomolar concentrations.
  • Selective for class I and class IIb HDACs.
  • Demonstrated anti-leukemia activity in preclinical models.
  • Available as a solid and as a 10 mM solution in DMSO.
  • High purity (approximately 99.5%).
  • Includes supporting documentation: data sheet, COA, SDS, H-NMR, and LC-MS.
Catalog No. 50-002-86225 Supplier Medchemexpress LLC Supplier No. HY15010910MG
Add to Cart
Add to Cart
Purinostat mesylate is the mesylate salt of purinostat, a potent and selective histone deacetylase (HDAC) inhibitor used in preclinical research. It exhibits low-nanomolar potency against multiple HDAC isoforms and has demonstrated apoptosis induction and cell-cycle effects in cancer models. The product is supplied as a solid or as a ready-to-use DMSO solution for laboratory studies.
  • Potent HDAC inhibition at low-nanomolar concentrations.
  • Selective for class I and class IIb HDACs.
  • Demonstrated anti-leukemia activity in preclinical models.
  • Available as a solid and as a 10 mM solution in DMSO.
  • High purity (approximately 99.5%).
  • Includes supporting documentation: data sheet, COA, SDS, H-NMR, and LC-MS.