Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Puritan Cary-Blair Medium

Catalog No. 22029646
Change view
Click to view available options
Fill Volume:
2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
22-029-646 5 mL
22-029-645 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-029-646 Supplier Puritan™ Supplier No. CB500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in the preservation of clinical fecal and rectal specimens containing enteric bacteria from the collection site to the testing laboratory for examination and culture.

  • 5 ml Cary-Blair Medium16 x 100 mm screw-cap polypropylene tube, orange capIntended for use in the preservation of clinical fecal and rectal specimens containing enteric bacteria from the collection site to the testing laboratory for examination and culture.
  • Premium medical grade plastic components to improve product shelf life.For optimum performance, store at 2-25°C.

Specifications

Appearance Characteristic
Broth Type Cary Blair Medium
Cap Color Orange
Cap Type Screw
Certifications/Compliance Yes
Color Clear Tube
Content And Storage 2 to 25°C
Description Cary Blair Medium
Detects Enteric bacteria
Format Tubes
Manufacturing Origin United States
Media Type Cary Blair Medium
Product Type Prepared Microbiology Tubes
Sterility Sterile
Test Type Preservation Efficacy Testing
Fill Volume 5 mL
Formulation Sodium Chloride 0.3%, Calcium Chloride 3.00%, Sodium Thioglycolate 0.15%, L-Cysteine 0.1%
For Use With (Application) Preservation of Clinical Fecal and Rectal Specimens
Material Polypropylene
No. per Pack 50
Packaging Type 50 Bulk
Quantity 50
Dimensions (Dia. x H) 16 x 100 mm
Height (Metric) 100 mm
Diameter (Metric) 16 mm
Manufacturing Quality CE IVD, Compliant with Regulation (EU) 2023/607 and Council Directive 93/42/EEC (MDD)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.