Learn More
Puritan™ Cotton-Tipped Plastic-Shaft Sterile Applicators
Catalog No. 22029499
Change view
Click to view available options
Quantity:
Individually wrapped
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-029-499 Pack of 2
22-029-504 Individually wrapped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-029-499 Supplier Puritan™ Supplier No. 25806 2PC
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Cotton-tipped sterile applicators with 6 in. plastic shafts
All purpose general swab; Wound dressing and cleaning; Application of topical dressings; Specimen collection; Cell collection for DNA testing; Individually wrapped, 1 or 2 per/package; Made in USA
Specifications
Quantity Pack of 2
Length (English) 6 in.
For Use With (Application) All purpose general swab, wound care, specimen collection
Product Type Applicator
Length (Metric) 15.2 cm
Sterility Sterile
Handle Material Polystyrene
Tip Style Regular
Tip Material Cotton