Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Puritan Lim Broth Medium

Catalog No. 22029649
Change view
Click to view available options
Fill Volume:
2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
22-029-649 2 mL
22-029-650 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-029-649 Supplier Puritan™ Supplier No. LB200
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use in the qualitative procedures for the isolation of Group B Streptococcus (GBS) from clinical specimens as well as in vitro diagnostic use.

2ml Lim Broth with Colistin and Nalidixic Acid.  12 x 80mm screw-cap tube, light blue cap.  lective enrichment medium for use in the qualitative procedures for the isolation of Group B Streptococcus (GBS) from clinical specimens.  Selective growth promotion of GBS and inhibition of gram-negative organisms, such as Escherichia coli and Proteus mirabilis.  For in Vitro Diagnostic Use.  Premium medical grade plastic components to improve product shelf life.  For optimum performance, store at 2-25°C.  Made in USA

Specifications

Broth Type Lim Broth
Cap Color Light Blue
Cap Type Screw
Certifications/Compliance Yes
Color Clear Tube
Content And Storage 2 to 25°C
Description Lim Broth Medium
Format Tube
Manufacturing Origin United States
Media Type Lim Broth
Sterility Sterile
Fill Volume 2 mL
Incubation Conditions Incubate tube aerobically or in 5% CO2 at 35-37°C.
Incubation Time 18-24 Hours
Packaging Type 50 Bulk
Quantity 50
Dimensions (Dia. x H) 12 x 80 mm
Height (Metric) 80 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.