Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Ayers Style Wooden Scraper

Catalog No. 1437210
Change view
Click to view available options
Quantity:
500 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-372-10 500 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-372-10 Supplier Puritan™ Supplier No. 2173

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puritan7 in. Ayers Style Wooden Scraper is naturally biodegradable and has splinter free edges and can be used as a general wooden scraper.

Splinter free edges. Naturally biodegradable. Made in USA

Specifications

Product Type Scraper
Material Wood
Sterility Non-sterile
Style Ayers
Length (English) 7 in.
Length (Metric) 17.7 cm
For Use With (Application) Scraping and collection of cervical cells for pap smears
Quantity 500 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.