Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ UniTranz-RT Transport Systems

Catalog No. 22027101
Change view
Click to view available options
Fill Volume:
1 mL
3 mL
Swab Configuration:
Dual Applicator
Single Applicator
Shaft Material:
Aluminum, Plastic
Polystyrene
Tip Type:
1 Standard, 1 Minitip
Elongated Flocked
Elongated, Ultrafine Flocked
Minitip Flocked
Ultrafine Flocked
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume Swab Configuration Shaft Material Tip Type
22-027-101 1 mL Single Applicator Polystyrene Minitip Flocked
22-027-103 3 mL Single Applicator Polystyrene Elongated Flocked
22-027-102 1 mL Single Applicator Polystyrene Ultrafine Flocked
22-027-105 3 mL Single Applicator Polystyrene Ultrafine Flocked
22-027-104 3 mL Single Applicator Polystyrene Minitip Flocked
22-027-108 3 mL Dual Applicator Aluminum, Plastic 1 Standard, 1 Minitip
22-027-106 3 mL Dual Applicator Polystyrene Elongated, Ultrafine Flocked
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 22-027-101 Supplier Puritan™ Supplier No. UT116
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma

Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination;Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Used in viral antigen detection test, PCR and rapid tests; Available in a 1ml or 3ml fill, medium is stable at room temperature; Advanced kit design is easy to use and reliable; Self-centering cap is leakproof and facilitates easy swab removal and disposal; High-performance swab tip features patented PurFlock Ultra™ swab with molded breakpoint designed to accommodate different collection sites; Soft tip for patient comfort; Premium medical grade plastic components to improve product shelf life; All kits include sterile wrapped swabs and vial; Shelf life - 18 months; Made in USA

Specifications

Product Type UniTranz-RT Transport System
Media Type Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
Sterility Sterile
Swab Configuration Single Applicator
Certifications/Compliance CE
Fill Volume 1 mL
For Use With (Application) Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Includes High performance mini tip purFlock ultra swab, wrapped and 1mL Universal Transport Medium Filled Vial
Packaging Individually-wrapped
pH 7.3 ±0.2
Quantity 300 Cs.
Shaft Material Polystyrene
Storage Requirements 2°C to 25°C
Tip Type Minitip Flocked
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.