Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Puritan™ Puritan MRSA Transport Medium, 2 ml

Catalog No. 22029647
Change view
Click to view available options
Fill Volume:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume
22-029-647 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-029-647 Supplier Puritan™ Supplier No. MRSA200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MRSA Transport Medium is an enrichment medium for the isolation of MRSA specimens

  • 2 ml MRSA Transport Medium.
  • 12 x 80 mm screw-cap polypropylene tube, pink cap.
  • Enrichment broth medium used for the isolation of specimens containing Staphylococcus aureusspecies, particularly methicillin-resistant Staphylococcus aureus (MRSA).
  • For In Vitro Diagnostic Use.
  • Premium medical grade plastic components to improve product shelf life. For optimum performance, store at 2-25°C.

Specifications

Appearance Characteristic
Broth Type MRSA Transport Medium
Cap Color Pink Cap
Cap Type Screw
Certifications/Compliance Yes
Color Clear Tube
Concentration 2 mL
Content And Storage 2 to 25°C
Description MRSA Transport Medium
Additive Sodium Chloride
Detects Staphylococcus aureus Species, Methicillin-resistant Staphylococcus aureus (MRSA)
Format Tubes
Manufacturing Origin United States
Media Type Enrichment Broth Medium
Product Type Broth Tube
Sterility Sterile
Target Organism Class Bacteria
Test Type Manual Diagnostics
Fill Volume 2 mL
Formulation Enrichment Broth Medium
For Use With (Application) Isolation of MRSA \Specimens, In Vitro Diagnostic Use
Includes 2 mL MRSA Transport Medium, 12 x 80 mm Screw-cap Polypropylene Tube with a Pink Cap
Incubation Conditions Incubate inoculated MRSA transport medium at 35 ±2°C.
Incubation Time Examine the MRSA transport medium for growth after 18-24 hours.
Material Polypropylene
No. per Pack 50
Packaging Type 50 Bulk
Quantity 50
Dimensions (Dia. x H) 12 x 80 mm
Dimensions (L x W) 12 x 80 mm
Height (Metric) 80 mm
Diameter (Metric) 12 mm
Manufacturing Quality CE IVD
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.