Learn More
HYDROFLEX AMERICA INC PurMop EF40-S Sterile Disposable Cleanroom Mop, Case of 100
Catalog No. NC1673445
Encompass_Preferred
PurMop EF40-S Sterile Disposable Cleanroom Mop 100% Polyester Microfiber pile for high cleaning performance. Cleanroom mop with pockets, 40 cm system .Highest cleaning ability due to special microfiber technology. High area performance through multiple fluid absorbing inserts. Even and fine fluid distribution on the surface. Ergonomic handling through ideal gliding properties. Packaged 5 pcs/pack, 20 packs/case = 100 pcs/case
Catalog No. NC1673445 Supplier HYDROFLEX AMERICA INC Supplier No. 2111109
PurMop EF40-S Sterile Disposable Cleanroom Mop 100% Polyester Microfiber pile for high cleaning performance. Cleanroom mop with pockets, 40 cm system .Highest cleaning ability due to special microfiber technology. High area performance through multiple fluid absorbing inserts. Even and fine fluid distribution on the surface. Ergonomic handling through ideal gliding properties. Packaged 5 pcs/pack, 20 packs/case = 100 pcs/case