Learn More
Cayman Chemical Purmorphamine, 483367-10-8, 1 mg
SDP
Supplier:  Cayman Chemical 100096341
Encompass_Preferred
Molecular Formula C31H32N6O2, Purity ≥98%, Formula Weight 520.6; A 2,6,9-trisubstituted purine that promotes the differentiation of both human and mouse mesenchymal progenitor cells into osteoblasts; binds to and activates the 7-transmembrane Smo receptor of the Hedgehog signaling pathway.
Catalog No. NC9630435