Learn More
Apexbio Technology LLC Purmorphamine 483367-10-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062671
Encompass_Preferred
Purmorphamine (CAS 483367-10-8) is a synthetic small molecule agonist targeting the Smoothened (Smo) protein a critical component of the Hedgehog (Hh) signaling pathway By binding and activating Smo Purmorphamine induces downstream modulation of key pathway targets such as Gli1 and Patched In vitro studies demonstrated activation with an IC50 value of approximately 1 5 M during competitive assays with the antagonist cyclopamine and an EC50 of about 1 M for alkaline phosphatase (ALP) expression in C3H10T1/2 multipotent cells This agent has been applied in research contexts for inducing osteogenic differentiation with implications for studying bone regeneration and neural degeneration processes
Catalog No. 50-000-62671 Supplier Apexbio Technology LLC Supplier No. A82285.1
Purmorphamine (CAS 483367-10-8) is a synthetic small molecule agonist targeting the Smoothened (Smo) protein a critical component of the Hedgehog (Hh) signaling pathway By binding and activating Smo Purmorphamine induces downstream modulation of key pathway targets such as Gli1 and Patched In vitro studies demonstrated activation with an IC50 value of approximately 1 5 M during competitive assays with the antagonist cyclopamine and an EC50 of about 1 M for alkaline phosphatase (ALP) expression in C3H10T1/2 multipotent cells This agent has been applied in research contexts for inducing osteogenic differentiation with implications for studying bone regeneration and neural degeneration processes