Learn More
Enzo Life Sciences Purmorphamine (25mg). CAS: 483367-10-8
SDP
Supplier:  Enzo Life Sciences ALX420045M025
Encompass_Preferred
Cell permeable purine compound. Induces osteoblast differentiation. Agonist of smoothened by activation of the Hedgehog (Hh) pathway. Also shown to induce transdifferentiation in pre-adipocytes and myoblasts. Binds to and activates 7-transmembrane receptor of Hh. Alternative name: 2-(1-Naphthoxy)-6-(4-morpholinoanilino)-9-cyclohexylpurine. Purity: ≥98% (HPLC). Appearance: Off-white to beige solid. Solubility: Soluble in DMSO (10mM). Long Term Storage: -20°C.
Catalog No. 50-200-9125