Learn More
Medchemexpress LLC Purmorphamine | 483367-10-8 | C31H32N6O2 | 250 MG
SDP
Catalog No. 5000374323
Encompass_Preferred
Purmorphamine is a smoothened/Smo receptor agonist with an EC50 of 1 μM. This compound, appearing as a white to off-white solid, has a molecular weight of 520.62 and a chemical formula of C31H32N6O2. It induces up-regulation of alkaline phosphatase activity, RUNX-2, and osteocalcin, and also induces STAT3 phosphorylation in mouse ES cell line ES14 and mesenchymal stem cell line C3H10T1/2.
  • Acts as a smoothened/Smo receptor agonist
  • Induces up-regulation of alkaline phosphatase activity and RUNX-2 expression
  • Promotes up-regulation of osteocalcin
  • Induces STAT3 phosphorylation in mouse ES cell line ES14 and mesenchymal stem cell line C3H10T1/2
  • Decreases cell numbers when combined with sirolimus
  • Up-regulates markers of the osteoblast phenotype in human bone marrow mesenchymal cells
  • Cited in studies for various assays including CCK-8, cell counting, and in vivo efficacy
Catalog No. 50-003-74323 Supplier Medchemexpress LLC Supplier No. 250MGHY15108
Purmorphamine is a smoothened/Smo receptor agonist with an EC50 of 1 μM. This compound, appearing as a white to off-white solid, has a molecular weight of 520.62 and a chemical formula of C31H32N6O2. It induces up-regulation of alkaline phosphatase activity, RUNX-2, and osteocalcin, and also induces STAT3 phosphorylation in mouse ES cell line ES14 and mesenchymal stem cell line C3H10T1/2.
  • Acts as a smoothened/Smo receptor agonist
  • Induces up-regulation of alkaline phosphatase activity and RUNX-2 expression
  • Promotes up-regulation of osteocalcin
  • Induces STAT3 phosphorylation in mouse ES cell line ES14 and mesenchymal stem cell line C3H10T1/2
  • Decreases cell numbers when combined with sirolimus
  • Up-regulates markers of the osteoblast phenotype in human bone marrow mesenchymal cells
  • Cited in studies for various assays including CCK-8, cell counting, and in vivo efficacy