Learn More
Selleck Chemical LLC Purmorphamine 5mg 483367-10-8
SDP
Supplier:  Selleck Chemical LLC S30425MG
Encompass_Preferred
Purmorphamine, which directly binds and activates Smoothened, blocks BODIPY-cyclopamine binding to Smo with IC50 of ~ 1.5 ?M in HEK293T cell and also is an inducer of osteoblast differentiation with EC50 of 1 ?M. Purmorphamine can reduce both basal and induced autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3303