Learn More
Apexbio Technology LLC Purmorphamine, 5mg. Cas: 483367-10-8 MFCD: MFCD09037557. Hedgehog agonist
SDP
Supplier:  Apexbio Technology LLC A82285
Encompass_Preferred
Purmorphamine is the first small-molecule agonist developed for the protein Smoothened [1]. Purmorphamine activates the Hedgehog (Hh) signaling pathway, resulting in the up- and downregulation of its downstream target genes, including Gli1 and Patched. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5083