Learn More
Cayman Chemical Purmorphamine Other Receptor M
Supplier: Cayman Chemical 100096345 MG
A 2,6,9-trisubstituted purine that promotes the differentiation of both human and mouse mesenchymal progenitor cells into osteoblasts; binds to and activates the 7-transmembrane Smo receptor of the Hedgehog signaling pathway