Learn More
Cayman Chemical Purmorphamine Other Receptor M
SDP
Catalog No. 502887186
Encompass_Preferred
A 2,6,9-trisubstituted purine that promotes the differentiation of both human and mouse mesenchymal progenitor cells into osteoblasts; binds to and activates the 7-transmembrane Smo receptor of the Hedgehog signaling pathway
Catalog No. 50-288-7186 Supplier Cayman Chemical Supplier No. 100096341 MG
A 2,6,9-trisubstituted purine that promotes the differentiation of both human and mouse mesenchymal progenitor cells into osteoblasts; binds to and activates the 7-transmembrane Smo receptor of the Hedgehog signaling pathway