Learn More
Bioworld Purolite Mixed Bed Resin UCW3700, 100 g
Catalog No. 50197473
Encompass_Preferred
Purolite Mixed Bed ResinMixed bed resins are used in the production of ultra pure water, preparation of condensate, radiation waste treatment and in the manufacture and purification of pharmaceutical products. Purolite mixed bed resins are strong base type 1 anion exchanger. Mixed bed resins can be used for both regenerable or non-regenerable (cartridge) systems.Basic ApplicationPolymer Structure : Gel polystyrene crosslinked with divinylbenzeneAppearance : Spherical beadsFunctional Group : Sulphonic Acid and Type 1 Quaternary AmmoniumIonic form as shipped : H+ / OH-
Catalog No. 50-197-473 Supplier Bioworld Supplier No. 201000051
Purolite Mixed Bed ResinMixed bed resins are used in the production of ultra pure water, preparation of condensate, radiation waste treatment and in the manufacture and purification of pharmaceutical products. Purolite mixed bed resins are strong base type 1 anion exchanger. Mixed bed resins can be used for both regenerable or non-regenerable (cartridge) systems.Basic ApplicationPolymer Structure : Gel polystyrene crosslinked with divinylbenzeneAppearance : Spherical beadsFunctional Group : Sulphonic Acid and Type 1 Quaternary AmmoniumIonic form as shipped : H+ / OH-