Learn More
Invivogen Puromycin, Selective antibiotic for the Pac gene, 58-58-2, 100mg (10x1ml)
SDP
Catalog No. NC9138068
Encompass
Formula: C22H29N7O5 • 2HCl, Purity: ≥ 98% (HPLC), Molecular weight: 544.3, Endotoxin level: < 5 EU/mg, Puromycin is an aminonucleoside antibiotic produced by Streptomyces alboniger. It specifically inhibits peptidyl transfer on both prokaryotic and eukaryotic ribosomes. This antibiotic inhibits the growth of Gram positive bacteria and various animal and insect cells.
Catalog No. NC9138068 Supplier Invivogen Supplier No. ANTPR1
Formula: C22H29N7O5 • 2HCl, Purity: ≥ 98% (HPLC), Molecular weight: 544.3, Endotoxin level: < 5 EU/mg, Puromycin is an aminonucleoside antibiotic produced by Streptomyces alboniger. It specifically inhibits peptidyl transfer on both prokaryotic and eukaryotic ribosomes. This antibiotic inhibits the growth of Gram positive bacteria and various animal and insect cells.