Learn More
Selleck Chemical LLC Puromycin 2HCl
SDP
Catalog No. 5000001493
Encompass_Preferred
Puromycin 2HCl is an aminonucleoside antibiotic derived from Streptomyces alboniger It is an inhibitor of protein synthesis and has been used to study transcription regulatory mechanisms that control the sequential and coordinated expression of genes during cell differentiation
Catalog No. 50-000-01493 Supplier Selleck Chemical LLC Supplier No. S7417500MG
Puromycin 2HCl is an aminonucleoside antibiotic derived from Streptomyces alboniger It is an inhibitor of protein synthesis and has been used to study transcription regulatory mechanisms that control the sequential and coordinated expression of genes during cell differentiation