Learn More
Selleck Chemical LLC Puromycin 2HCl, 25 mg
SDP
Supplier:  Selleck Chemical LLC S741725MG
Encompass_Preferred
Puromycin 2HCl is an aminonucleoside antibiotic derived from Streptomyces alboniger. It is an inhibitor of protein synthesis and has been used to study transcription regulatory mechanisms that control the sequential and coordinated expression of genes during cell differentiation.
Catalog No. NC2853813